Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin R1/SSTR1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227413SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227413SS 0.025 mg
NBP227413 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227413SS Supplier Novus Biologicals Supplier No. NBP227413SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Somatostatin R1/SSTR1 Polyclonal specifically detects Somatostatin R1/SSTR1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Somatostatin R1/SSTR1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias G-protein coupled receptor, somatostatin receptor 1, somatostatin receptor type 1, SRIF-2, SS1R, SS-1-R, SS1-R, SSR1
Gene Symbols SSTR1
Host Species Rabbit
Immunogen This antibody was raised against Somatostatin Receptor 1
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6751
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.