Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Somatostatin R1/SSTR1 Antibody (SR2064), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32195750UL
Description
Somatostatin R1/SSTR1 Monoclonal antibody specifically detects Somatostatin R1/SSTR1 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Somatostatin R1/SSTR1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
SR2064 | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
G-protein coupled receptor, somatostatin receptor 1, somatostatin receptor type 1, SRIF-2, SS1R, SS-1-R, SS1-R, SSR1 | |
A synthesized peptide derived from human Somatostatin R1/SSTR1 (Uniprot #: P30872) | |
50 μg | |
Breast Cancer, Cancer, Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
6751 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction