Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Somatostatin R2/SSTR2 Antibody (7A9), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Somatostatin R2/SSTR2 |
---|---|
Clone | 7A9 |
Dilution | Flow Cytometry, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
Somatostatin R2/SSTR2 Monoclonal antibody specifically detects Somatostatin R2/SSTR2 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
Somatostatin R2/SSTR2 | |
Flow Cytometry, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
somatostatin receptor 2, somatostatin receptor type 2, SRIF-1, SS2R, SS-2-R, SS2-R | |
Recombinant Human Somatostatin R2/SSTR2 protein [UniProt P30874] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
7A9 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Human | |
Alzheimers Research, Cancer, Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
6752 | |
IgG1 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title