Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Somatostatin R3/SSTR3 Antibody (576017), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7018PECY55
Description
Somatostatin R3/SSTR3 Monoclonal antibody specifically detects Somatostatin R3/SSTR3 in Human samples. It is validated for Flow CytometrySpecifications
Somatostatin R3/SSTR3 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
576017 | |
Flow Cytometry | |
somatostatin receptor 3, SS-3-R, SS3-R | |
NS0 mouse myeloma cell line transfected with human Somatostatin R3/SSTR3, Accession # P32745 | |
0.1 mL | |
Apoptosis, Breast Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
6753 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction