Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Somatostatin R4/SSTR4 Antibody (OTI2F11), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274283AF700
Description
Somatostatin R4/SSTR4 Monoclonal specifically detects Somatostatin R4/SSTR4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Somatostatin R4/SSTR4 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SSTR4 | |
Synthetic peptide corresponding to residues near C-terminus of human Somatostatin R4/SSTR4 (NP_001043). | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI2F11 | |
Western Blot | |
G-protein coupled receptor, somatostatin receptor 4, somatostatin receptor type 4, SS4R, SS-4-R, SS4-R | |
Mouse | |
Affinity Purified | |
Breast Cancer, Cell Cycle and Replication | |
6754.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction