Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin R5/SSTR5 Antibody (394401), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB370091 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370091 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370091 Supplier Novus Biologicals Supplier No. FAB4448W
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Somatostatin R5/SSTR5 Monoclonal antibody specifically detects Somatostatin R5/SSTR5 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Somatostatin R5/SSTR5
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 394401
Conjugate DyLight 650
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias somatostatin receptor 5, somatostatin receptor subtype 5, somatostatin receptor type 5, SS5R, SS-5-R, SS5-R
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Somatostatin R5/SSTR5, Met1-Leu364, Accession # AAK61266
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6755
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.