Learn More
SOMNI Scientific™ SOMNI Scientific™ SOMNI Bain Circuit
SDP
Catalog No. 04777210 Shop All Somni Scientific Products
Click to view available options
Length (English):
60 in.
72 in.
The tube within a tube design provides heat and moisture conservation with fresh gas entering the circuit at the patient end. GS elbow included. PVC, Phthalate and Latex Free
  • A tube within a tube design that provides heat and moisture conservation
  • The fresh gas enters the circuit at the patient end
  • PVC, Phthalate and Latex Free
  • GS elbow included
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
Length (English) 72 in.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.