Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SON Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595740
Description
Positive Samples: LO2, HeLa Immunogen sequence: TMDSQMLATS SMDSQMLASG TMDSQMLASG TMDAQMLASG TMDAQMLASS TQDSAMLGSK SPDPYRLAQD PYRLAQDPYR LGHDPYRLGH DAYRLGQDPY R.
The protein encoded by this gene binds to a specific DNA sequence upstream of the upstream regulatory sequence of the core promoter and second enhancer of human hepatitis B virus (HBV). Through this binding, it represses HBV core promoter activity, transcription of HBV genes, and production of HBV virions. The protein shows sequence similarities with other DNA-binding structural proteins such as gallin, oncoproteins of the MYC family, and the oncoprotein MOS. It may also be involved in protecting cells from apoptosis and in pre-mRNA splicing. Several transcript variants encoding different isoforms have been described for this gene, but the full-length nature of only two of them has been determined. [provided by RefSeq].
Specifications
SON | |
Polyclonal | |
Unconjugated | |
SON | |
2900011L12Rik; AA409051; AU067731; BASS1; Bax antagonist selected in Saccharomyces 1; C21orf50; C81487; DBP5; DBP-5; HSPC310; HSPC312; KIAA1019; mKIAA1019; negative regulatory element-binding protein; NRE-binding protein; NREBP; Protein DBP-5; Protein SON; SON; Son cell proliferation protein; Son DNA binding protein; SON DNA-binding protein; SON3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6651 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.18 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P18583 | |
SON | |
A synthetic peptide corresponding to a sequence within amino acids 850-950 of human SON (NP_0012783401). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction