Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Sonelokimab Humanized Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542290
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Sonelokimab is a biosimilar that targets ALB,IL17A.
Specifications
Sonelokimab Humanized | |
Recombinant Monoclonal | |
Unconjugated | |
M-1095; MSB-0010841 | |
Human ALB,IL17A. | |
100 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA | |
1.42 mg/mL | |
30mM histidine with 10% sucrose, 0.02% Tween 80 and no preservative; pH 5.8 | |
Human | |
Metal-chelate chromatography | |
RUO | |
Human | |
Antibody | |
Chemical |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction