Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sonic Hedgehog/Shh Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222139AF750
Description
Sonic Hedgehog/Shh Polyclonal specifically detects Sonic Hedgehog/Shh in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Sonic Hedgehog/Shh | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
HHG1, HHG-1, HLP3, HPE3, MCOPCB5sonic hedgehog (Drosophila) homolog, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SHH | |
A synthetic peptide made to an N-terminal portion of the human SHH protein (between residues 1-75) [UniProt Q15465] | |
0.1 mL | |
Biologically Active Proteins, Neuroscience, Sensory Systems, Stem Cell Signaling Pathway, Stem Cells, Vision | |
Human, Mouse, Rat, Primate | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction