Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Sonidegib is a potent Smoothened (Smo) receptor antagonist (IC50 values are 1.3 nM and 2.5 nM at mouse and human receptors, respectively). Inhibits growth of a variety of tumor types including basal cell carcinoma, small cell lung cancer and medulloblastoma. Sonidegib inhibits Hh pathway in CD34+ chronic myeloid leukemia cells; it reduces both the number of cells and self-renewal ability. Brain penetrant and orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | rel-N-[6-[(2R,6S)-2,6-Dimethyl-4-morpholinyl]-3-pyridinyl]-2-methyl-4'-(trifluoromethoxy)[1,1'-biphenyl]-3-carboxamide |
| CAS | 956697-53-3 |
| Quantity | 50 mg |
| Target | Smoothened |
| Molecular Formula | C26H26F3N3O3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

