Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sonoco™ ThermoSafe Certis™ Gold Shipper Box with Universal Packout

Catalog No. p-4903569 Shop All Sonoco ThermoSafe Products
Change view
Click to view available options
Duration:
126 hrs
78 hr.
Material:
Polyurethane
VIP
Payload:
40 lb.
80 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Duration Material Payload
03-531-201 78 hr. Polyurethane 40 lb.
03-531-204 126 hrs VIP 80 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-531-201 Supplier Sonoco™ Supplier No. CRUG3XP
Only null left
Add to Cart
Add to Cart
Request a Quote

Accommodates broad range of temperature-sensitive products

With flexible, modular design, these shippers can adjust to meet customer requirements for temperature control, shipping durations and payload sizes. ThermoPhase™ powered by PureTemp™ non-petroleum phase-change technology enables strict temperature control, easy pack outs, greater efficiency and packages that are smaller and lighter. Three packout options include seasonal and year-round temperature protection.

  • Choice of two ambient profiles (ISCgold™ and ISCsilver™)
  • Easily modified to align with product specifications
  • Off-the-shelf convenience
  • Reduced shipping costs and transit time to market

Certis Gold:

  • ISCgold ambient profile to meet in-transit temperature extremes for drug products
  • Three to five day durations
  • Universal packout

TRUSTED_SUSTAINABILITY

Specifications

Applications Accommodates broad range of temperature-sensitive products
Color Gold
Duration 78 hr.
Payload 40 lb.
Material Polyurethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.