Learn More
Sonoco™ ThermoSafe Insulated Foam Shipper Available on GSA/VA Contract for Federal Government customers only.
EPS foam-only shippers offer the ideal solution for the safe storage and delivery of most refrigerated or frozen products
Manufacturer:  Sonoco™ 481
 View more versions of this product
Catalog No. 03-001-216
  Check Availability
Add to cart
Description
Description
  • Large selection of cost-effective, reusable stock containers available
  • Excellent dry ice sublimation – average dry ice usage in 1 1/2 in. thick wall units is 5 pounds over 24 hours, and 3-4 pounds thereafter
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
42 in.
106.6cm
23 in.
58.4cm
19 in.
48.2cm
Safe storage and delivery of most refrigerated or frozen products
ID: 19 x 14.625 x 12.25 in.
Insulated
12.25 in.
31.1cm
19 in.
48.2cm
14.6 in.
37cm
3/Cs.
23 x 19 x 42 in. (58.4 x 48.2 x 106.6cm)