Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbic Acid, Powder, NF, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18606917 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle

SO215, 110-44-1, C6H8O2

Spectrum™ Chemical Sorbic Acid, Powder, NF is used as an antimicrobial agent in pharmaceutical formulations. Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 110-44-1
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00002703
InChI Key WSWCOQWTEOXDQX-MQQKCMAXSA-N
IUPAC Name (2E,4E)-hexa-2,4-dienoic acid
SMILES C\C=C\C=C\C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.002
Melting Range 132-135°C
Packaging Amber Glass Bottle
Quantity 2.5 kg
Percent Purity 99 to 101%
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Sorbic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.