Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitan monolaurate

Catalog No. p-7045821
Click to view available options
Quantity:
250 g
1000 g

CAS: 1338-39-2 | C18H34O6 | 346.46 g/mol

Sorbitan monolaurate is a detergent and monoester of lauric acid and hexitol anhydrides derived from sorbitol. It is used in a study to assess enhanced solubility and oral bioavailability of itraconazole. It is also used in a study to investigate the oxidation of unsaturated lipids in oil-in-water emulsions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sorbitan monolaurate is a detergent and monoester of lauric acid and hexitol anhydrides derived from sorbitol. It is used in a study to assess enhanced solubility and oral bioavailability of itraconazole. It is also used in a study to investigate the oxidation of unsaturated lipids in oil-in-water emulsions.

Solubility
Soluble in 2-ethoxyethanol, ethanol and methanol. Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1338-39-2
Molecular Formula C18H34O6
Molecular Weight (g/mol) 346.46
MDL Number MFCD00005365
InChI Key LWZFANDGMFTDAV-IOVMHBDKSA-N
Synonym Sorbitan monododecanoate; Span™ 20
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

Density 1.032
Quantity 250 g
Flash Point >110°C (230°F)
Refractive Index 1.474
Merck Index 14,8724
Solubility Information Soluble in 2-ethoxyethanol,ethanol and methanol. Not miscible or difficult to mix in water.
Formula Weight 346.47
Chemical Name or Material Sorbitan monolaurate
EINECSNumber 215-663-3
RTECSNumber WG2920000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.