Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitol Dehydrogenase Antibody (10F4), FITC, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242653F

Catalog No. NBP242653F


Only null left
Add to Cart

Description

Description

Sorbitol Dehydrogenase Monoclonal specifically detects Sorbitol Dehydrogenase in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Sorbitol Dehydrogenase
Monoclonal
FITC
PBS with 0.05% Sodium Azide
SORD
Recombinant human SORD (1-357aa) purified from E. coli
100 μL
Diabetes Research, Lipid and Metabolism
6652
Store at 4C in the dark.
IgG1 κ
ELISA, Immunocytochemistry, Immunofluorescence
10F4
ELISA, Immunocytochemistry/Immunofluorescence
EC 1.1.1, EC 1.1.1.14, L-iditol 2-dehydrogenase, sorbitol dehydrogenase, SORD1
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.