Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sorbitol Dehydrogenase Antibody (OTI9E7), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274288FR
Description
Sorbitol Dehydrogenase Monoclonal specifically detects Sorbitol Dehydrogenase in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
Sorbitol Dehydrogenase | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
SORD | |
Full length recombinant protein of human Sorbitol Dehydrogenase (NP_003095) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI9E7 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
EC 1.1.1, EC 1.1.1.14, L-iditol 2-dehydrogenase, sorbitol dehydrogenase, SORD1 | |
Mouse | |
Affinity Purified | |
Diabetes Research, Lipid and Metabolism | |
6652.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction