Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sorbitol Dehydrogenase Monoclonal Antibody (OTI9E7)
GREENER_CHOICE

Catalog No. PIMA524995
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524995 Supplier Invitrogen™ Supplier No. MA524995
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sorbitol Dehydrogenase Monoclonal Antibody for Western Blot, ICC/IF, Flow

Sorbitol dehydrogenase catalyzes the interconversion of polyols and their corresponding ketoses, and together with aldose reductase is the reduction of glucose to sorbitol by ALDR1 with NADPH as the cofactor. SORD then oxidizes the sorbitol to fructose using NADcofactor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sorbitol Dehydrogenase
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI9E7
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SORD
Gene Accession No. Q00796
Gene Alias (R,R)-butanediol dehydrogenase; epididymis secretory sperm binding protein Li 95n; HEL-S-95n; L-iditol 2-dehydrogenase; LOW QUALITY PROTEIN: sorbitol dehydrogenase; polyol dehydrogenase; RDH; Ribitol dehydrogenase; SDH; Sdh1; Sdh-1; Sodh-1; Sorbitol dehydrogenase; sorbitol dehydrogenase 1; SORD; SORD 1; SORD1; XDH; Xylitol dehydrogenase
Gene Symbols SORD
Host Species Mouse
Immunogen Full length human recombinant protein of SORD produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6652
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.