Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SORBS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569798
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569798 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569798 Supplier Invitrogen™ Supplier No. PA569798
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 92%.

Plays a role in tyrosine phosphorylation of CBL by linking CBL to the insulin receptor. Required for insulin-stimulated glucose transport. Involved in formation of actin stress fibers and focal adhesions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SORBS1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SORBS1
Gene Accession No. Q62417
Gene Alias 2310065E01Rik; 9530001P15Rik; CAP; c-Cbl associated protein; c-Cbl-associated protein; DKFZp451C066; DKFZP586P1422; Fas-ligand associated factor 2; FLAF2; FLJ12406; I79_015895; KIAA0894; KIAA1296; mKIAA1296; OTTHUMP00000020142; OTTHUMP00000020143; OTTHUMP00000020144; OTTHUMP00000020145; Ponsin; R85FL; SH3 domain protein 5; Sh3d5; SH3P12; SORB1; sorbin and SH3 domain containing 1; sorbin and SH3 domain-containing protein 1; Sorbs1; sorbs1 {ECO:0000250; UniProtKB:Q62417}
Gene Symbols SORBS1
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse SORBS1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20411
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.