Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SORBS2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986214
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6214 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6214 Supplier Bioss Supplier No. BS4905R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Arg and c-Abl represent the mammalian members of the Abelson family of non-receptor protein-tyrosine kinases. They interact with the Arg/Abl binding proteins via the SH3 domains present in the carboxy end of the latter group of proteins. ARGBP2 is the sorbin and SH3 domain containing 2 protein. It has three C-terminal SH3 domains and an N-terminal sorbin homology(SoHo) domain that interacts with lipid raft proteins. The subcellular localization of this protein in epithelial and cardiac muscle cells suggests that it functions as an adapter protein to assemble signaling complexes in stress fibers, and that it is a potential link between Abl family kinases and the actin cytoskeleton.

Specifications

Antigen SORBS2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SORBS2
Gene Accession No. O94875
Gene Alias 2010203O03Rik; 9430041O17Rik; A530071H08; Arg binding protein 2; Arg/Abl-interacting protein 2; Arg/Abl-interacting protein ArgBP2; Arg-binding protein 2; ARGBP2; BOS_23784; KIAA0777; mKIAA0777; nArgBP2; Neural ArgBP2; PRO0618; Sorbin; sorbin and SH3 domain containing 2; Sorbin and SH3 domain-containing protein 2; Sorbs2
Gene Symbols SORBS2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SORBS2/ArgBP2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8470
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.