Learn More
Invitrogen™ Sorcin Polyclonal Antibody

Description
Sequence homology: Xenopus (93%), Opossum (87%), Chicken (81%).
Specifications
Specifications
| Antigen | Sorcin |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Sri |
| Gene Accession No. | P30626, Q6P069 |
| Gene Alias | 22 kDa protein; 2210417O06Rik; 2900070H08Rik; calcium binding protein amplified in mutlidrug-resistant cells; CP22; CP-22; H_RG167B05.1; SCN; Sor; Sorcin; SRI; V19 |
| Gene Symbols | Sri |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.