Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SorCS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552883
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552883 Supplier Invitrogen™ Supplier No. PA552883
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EWRRDIGRVI KKSLVEATGV PGQHILVAVL PGLPTTAELF VLPYQDPAGE NKRSTDDLEQ ISELLIHTLN QNSVHFELKP GVRVLVHAAH LTAAPLVDLT Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.

SORCS1 is one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SorCS1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sorcs1
Gene Accession No. Q8WY21
Gene Alias FLJ41758; FLJ43475; FLJ44957; hSorCS; mSorCS; Sorcs; SORCS receptor 1; SORCS1; sortilin related VPS10 domain containing receptor 1; sortilin-related VPS10 domain containing receptor 1; VPS10 domain receptor protein SORCS 1; VPS10 domain-containing receptor SorCS1
Gene Symbols Sorcs1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SorCS1. Recombinant protein control fragment (Product #RP-90098).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114815
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.