Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SorCS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113257
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113257 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113257 Supplier Invitrogen™ Supplier No. PA5113257
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SorCS1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

SORCS1 is one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SorCS1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Sorcs1
Gene Accession No. Q8WY21
Gene Alias FLJ41758; FLJ43475; FLJ44957; hSorCS; mSorCS; Sorcs; SORCS receptor 1; SORCS1; sortilin related VPS10 domain containing receptor 1; sortilin-related VPS10 domain containing receptor 1; VPS10 domain receptor protein SORCS 1; VPS10 domain-containing receptor SorCS1
Gene Symbols Sorcs1
Host Species Rabbit
Immunogen Recombinant Human VPS10 domain-containing receptor SorCS1 protein (34-175AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114815
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.