Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SorCS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547596
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547596 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547596 Supplier Invitrogen™ Supplier No. PA547596
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) SorCS2 and less than 5% cross-reactivity with rhSorCS1 and recombinant mouse SorCS1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

This gene encodes one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein. Members of this gene family are large with many exons but the CDS lengths are usually less than 3700 nt. Very large introns typically separate the exons encoding the VPS10 domain; the remaining exons are separated by much smaller-sized introns. These genes are strongly expressed in the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SorCS2
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene SORCS2
Gene Accession No. Q9EPR5
Gene Alias KIAA1329; mKIAA1329; N28137; SORCS2; SorCS2 104 kDa chain; SorCS2 122 kDa chain; SorCS2 18 kDa chain; sortilin related VPS10 domain containing receptor 2; sortilin-related VPS10 domain containing receptor 2; VPS10 domain receptor protein; VPS10 domain receptor protein SORCS 2; VPS10 domain-containing receptor SorCS2
Gene Symbols SORCS2
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SorCS2 Ala118-Gly1078.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81840
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.