Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SorCS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145014 Supplier Invitrogen™ Supplier No. PA5145014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSMDMNGKPT NCKPPDCHLH LHLRWADNPY VSGTVHTKDT APGLIMGAGN LGSQLVEYKE EMYITSDCGH T Antigen sequence identity: Mouse 96%; Rat 97%.

This gene encodes one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein. Members of this gene family are large with many exons but the CDS lengths are usually less than 3700 nt. Very large introns typically separate the exons encoding the VPS10 domain; the remaining exons are separated by much smaller-sized introns. These genes are strongly expressed in the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SorCS2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SORCS2
Gene Accession No. Q96PQ0
Gene Alias KIAA1329; mKIAA1329; N28137; SORCS2; SorCS2 104 kDa chain; SorCS2 122 kDa chain; SorCS2 18 kDa chain; sortilin related VPS10 domain containing receptor 2; sortilin-related VPS10 domain containing receptor 2; VPS10 domain receptor protein; VPS10 domain receptor protein SORCS 2; VPS10 domain-containing receptor SorCS2
Gene Symbols SORCS2
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 483-553. Recombinant protein control fragment (Product #RP-109910).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57537
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.