Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SORCS2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981825
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1825 Supplier Bioss Supplier No. BS11963R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein. Members of this gene family are large with many exons but the CDS lengths are usually less than 3700 nt. Very large introns typically separate the exons encoding the VPS10 domain; the remaining exons are separated by much smaller-sized introns. These genes are strongly expressed in the central nervous system.

Specifications

Antigen SORCS2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SORCS2
Gene Accession No. Q96PQ0
Gene Alias KIAA1329; mKIAA1329; N28137; Sorcs2; SorCS2 104 kDa chain; SorCS2 122 kDa chain; SorCS2 18 kDa chain; sortilin related VPS10 domain containing receptor 2; sortilin-related VPS10 domain containing receptor 2; VPS10 domain receptor protein; VPS10 domain receptor protein SORCS 2; VPS10 domain-containing receptor SorCS2
Gene Symbols SORCS2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SORCS2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57537
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.