Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SorCS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548023
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548023 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548023 Supplier Invitrogen™ Supplier No. PA548023
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 40% cross-reactivity with recombinant human SorCS3 is observed, approximately 7% cross-reactivity with recombinant mouse (rm) SorCS1 is observed, and less than 2% cross-reactivity with rmSorCS2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SorCS3
Applications Immunohistochemistry (Frozen), Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene SORCS3
Gene Accession No. Q8VI51
Gene Alias 6330404A12Rik; AW045579; KIAA1059; SORCS; SORCS3; sortilin related VPS10 domain containing receptor 3; sortilin-related VPS10 domain containing receptor 3; VPS10 domain receptor protein SORCS 3 (SORCS3); VPS10 domain-containing receptor SorCS3
Gene Symbols SORCS3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SorCS3 Ala131-Ser1122.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66673
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.