Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SORCS3 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11503R

Specifications
SORCS3 | |
Polyclonal | |
Unconjugated | |
SORCS3 | |
6330404A12Rik; AW045579; SORCS; Sorcs3; sortilin-related VPS10 domain containing receptor 3; VPS10 domain-containing receptor SorCS3 | |
Rabbit | |
Protein A | |
RUO | |
22986 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9UPU3 | |
SORCS3 | |
KLH conjugated synthetic peptide derived from human SORCS3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction