Learn More
SORD Mouse anti-Human, Clone: OTI4E5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500749

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Sorbitol dehydrogenase catalyzes the interconversion of polyols and their corresponding ketoses, and together with aldose reductase is the reduction of glucose to sorbitol by ALDR1 with NADPH as the cofactor. SORD then oxidizes the sorbitol to fructose using NADcofactor.Specifications
SORD | |
Monoclonal | |
Unconjugated | |
SORD | |
HEL-S-95n, SORD1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6652 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI4E5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q00796 | |
SORD | |
Full length human recombit protein of human SORD produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG3 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.