Learn More
SORL1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305288AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rabbit
This gene encodes a mosaic protein that belongs to at least two families: the vacuolar protein sorting 10 domain-containing receptor family, and the low density lipoprotein receptor family. The encoded protein also contains fibronectin type III repeats and an epidermal growth factor repeat. The encoded protein is translated as a preproprotein and likely plays roles in endocytosis and sorting. There may be an association between expression of this locus and Alzheimer′s Disease.Specifications
SORL1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q92673 | |
SORL1 | |
Between 2164 and 2214 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SORL1 | |
SORL1; LR11; sortilin-related receptor, L(DLR class) A repeats-containing; LRP9; SORLA; gp250; SorLA-1; mosaic protein LR11; C11orf32; FLJ21930; FLJ39258; sortilin-related receptor containing LDLR class A repeats | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6653 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.