Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SORL1 Recombinant Rabbit Monoclonal Antibody (JE35-52)
GREENER_CHOICE

Catalog No. FEMA550824
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550824 Supplier Invitrogen™ Supplier No. MA550824
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: Jurkat cell lysates, mouse cerebellum tissue, rat cerebellum tissue. Predicted band size: 248 kDa Subcellular Location: Cell membrane, Cytoplasmic vesicle, Endoplasmic reticulum, Endosome, Golgi apparatus, Membrane, Secreted.

This gene encodes a mosaic protein that belongs to at least two families: the vacuolar protein sorting 10 domain-containing receptor family, and the low density lipoprotein receptor family. The encoded protein also contains fibronectin type III repeats and an epidermal growth factor repeat. The encoded protein is translated as a preproprotein and likely plays roles in endocytosis and sorting. There may be an association between expression of this locus and Alzheimer's Disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SORL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE35-52
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SORL1
Gene Accession No. O88307, Q92673
Gene Alias 2900010L19Rik; AI596264; AW261561; C11orf32; FLJ21930; FLJ39258; gp250; LDLR relative with 11 ligand-binding repeats; low-density lipoprotein receptor relative with 11 ligand-binding repeats; Lr11; LRP9; mosaic protein LR11; mSorLA; RGD619914; Sorl1; SorLA; SorLA-1; sortilin related receptor 1; sortilin-related receptor; sortilin-related receptor, L(DLR class) A repeats containing; sortilin-related receptor, LDLR class A repeats-containing; sorting protein-related receptor containing LDLR class A repeats
Gene Symbols SORL1
Host Species Rabbit
Immunogen Recombinant protein within Human SORL1 aa 1341-1540.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20660, 300652, 6653
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.