Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SorLA Antibody (SR1433), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | SorLA |
---|---|
Clone | SR1433 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
SorLA Monoclonal antibody specifically detects SorLA in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
SorLA | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
6653 | |
IgG | |
Affinity purified |
SR1433 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
C11orf32, chromosome 11 open reading frame 32, FLJ21930, FLJ39258, gp250, Low-density lipoprotein receptor relative with 11 ligand-binding repeats, LR11LDLR relative with 11 ligand-binding repeats, LRP9, mosaic protein LR11, SorLA, SorLA-1SORLA, sortilin-related receptor, sortilin-related receptor, L(DLR class) A repeats containing, sortilin-related receptor, L(DLR class) A repeats-containing, Sorting protein-related receptor containing LDLR class A repeats | |
A synthesized peptide derived from human SorLA (Uniprot #: Q92673) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title