Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Sortilin Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547462
Description
Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Sortilin functions as a sorting receptor in the Golgi compartment and as a clearance receptor on the cell surface. Sortilin is required for protein transport from the Golgi apparatus to the lysosomes by a pathway that is independent of the mannose-6-phosphate receptor (M6PR). It is also required for protein transport from the Golgi apparatus to the endosomes. Sortilin promotes neuronal apoptosis by mediating endocytosis of the proapoptotic precursor forms of BDNF (proBDNF) and NGFB (proNGFB). Also, Sortilin acts as a receptor for neurotensin.
Specifications
Sortilin | |
Polyclonal | |
Unconjugated | |
Sort1 | |
100 kDa NT receptor; 2900053A11Rik; AI852375; Glycoprotein 110; glycoprotein 95; Gp110; Gp95; LDLCQ6; mNTR3; neurotensin receptor 3; NT3; NTR3; Nts3; Ntsr3; Sort1; Sortilin; sortilin 1 | |
Goat | |
Antigen Affinity Chromatography | |
RUO | |
20661, 6272, 83576 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O54861, Q6PHU5, Q99523 | |
Sort1 | |
Mouse myeloma cell line NS0-derived recombinant human Sortilin Ser78-Asn755. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction