Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Sorting Nexin-11, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABC1444MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABC1444MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC1444MI Supplier MilliporeSigma™ Supplier No. ABC1444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sorting nexin-11 (UniProt Q91WL6) is encoded by the Snx11 gene (Gene ID 74479) in murine species. Sorting nexins (SNXs) are involved in diverse intracellular endosomal traffickings. Their phox homology (PX) domain binds certain phosphatidylinositols (PIs) and mediates their recruitment to PI-rich vesicles. Most SNXs contain additional recognizable domains and are referred to as SNX PX-BAR or SNX PX-other, while SNX3, SNX10, SNX11, SNX12, SNX22, and SNX24 contain only the PX domain and are referred to as SNX PX. SNX11 is found to contain a novel PXe domain (a.a. 16-132 of human and mouse SNX11) with two additional α-helices next to the conventional PX domain structure composed of three β-strands and three α-helices. F2RL1 (a.k.a. PAR2) is a protease-activated GPCR highly expressed throughout the central and the peripheral nervous system. F2RL1 is reported to translocate from the plasma membrane to nucleus upon stimulation via a microtubule-dependent shuttle machinery that requires sorting nexin 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sorting Nexin-11
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 0.05% Sodium Azide.
Gene Snx11
Gene Accession No. Q91WL6
Host Species Rabbit
Immunogen Linear peptide corresponding to an internal sequence from the C-terminal half of mouse sorting nexin-11.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001156861
Test Specificity This polyclonal antibody reacts with both human and murine sorting nexin-11. The immunogen sequence is also present in human spliced isoform 2 and rat sorting nexin-11.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.