Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Sorting Nexin-17, Mouse, Unlabeled, Clone: 4B8.1,

Catalog No. MABC1211MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABC1211MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1211MI Supplier MilliporeSigma™ Supplier No. MABC1211
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sorting nexin-17 (UniProt Q15036) is encoded by the SNX17 (also known as KIAA0064) gene (Gene ID 9784) in human. The sorting nexins (SNXs) constitute a family of adaptor proteins involved in proteins trafficking through the endosomal pathway. Originally identified as a binding partner of the cell adhesion protein P-selectin, sorting nexin-17 (SNX17) has since been shown to mediate internalization and recycling of multiple cell surface proteins including P-selectin, amyloid precursor protein (APP), integrin β1, low density lipoprotein receptor (LDLR), LDLR related protein (Lrp1), ApoER2, and FEEL1. All SNX proteins contain a conserved phox homology (PX) domain that mediates phosphoinositides binding in a SNX type-specific manner, as well as various scaffolding modules including SH3, BAR, and PDZ domains. SNX17 PX domain (a.a. 1-109) displays specificity for phosphatidylinositol 3-monophosphate, which is enriched at the cytoplasmic face of the early endosome. SNX17 also contains a 4.1 protein/ezrin/radixin/moesin (FERM) domain (a.a. 115-432) comprised of an ubiquitin/Ras association (RA) subdomain known as the F1 lobe (a.a. 113-205), an F2 lobe (a.a. 206-269) with a helical structure similar to acyl-CoA binding protein, and an F3 lobe (a.a. 270-388) similar to the phosphotyrosine binding (PTB)/pleckstrin homology domain. The FERM domain mediates interaction with Asn-Pro-Xaa-Tyr/Phe (NPX(Y/F) motif-containing proteins for internalization as well as interaction with adaptor proteins-containing PTB (or FERM) domains to trigger signaling events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sorting Nexin-17
Applications Western Blot
Classification Monoclonal
Clone 4B8.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene SNX17, KIAA0064
Gene Accession No. Q15036
Host Species Mouse
Immunogen GST-tagged recombinant fragment from the FERM-like region of human Sorting Nexin-17.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001253989
Test Specificity Clone 4B8.1 recognizes an internal epitope within the FERM-like region present in both spliced isoforms of human sorting nexin-17 reported by UniProt (Q15036).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.