Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorting Nexin 31 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen Sorting Nexin 31
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125211
SDP
View Documents
Novus Biologicals
NBP310155100UL
100 μg
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

Sorting Nexin 31 Polyclonal specifically detects Sorting Nexin 31 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Sorting Nexin 31
Western Blot
Unconjugated
Rabbit
Signal Transduction
PBS buffer, 2% sucrose
169166
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
Mouse
MGC39715, sorting nexin 31, sorting nexin-31
The immunogen is a synthetic peptide directed towards the middle terminal region of mouse Sorting Nexin 31 (NP_079988.3). Peptide sequence QIQDIAFQMSRVKCWQVTFLGTLLDTDGPQRTLNQNLELRFQYSEDSCQQ
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.