Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOS1 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP105954B
Description
SOS1 Polyclonal antibody specifically detects SOS1 in Human samples. It is validated for Western BlotSpecifications
SOS1 | |
Polyclonal | |
Western Blot | |
GF1GGF1, GINGF, guanine nucleotide exchange factor, HGFhereditary, 1, NS4, son of sevenless homolog 1, son of sevenless homolog 1 (Drosophila), SOS-1 | |
The immunogen recognized by this antibody maps to a region between residue 1025 and 1075 of human son of sevenless ortholog 1 using the numbering given in entry NP_005624.2 (GeneID 6654). | |
0.1 mL | |
Breast Cancer, Cancer | |
6654 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction