Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOS1 Antibody (SOS-01), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11068800AF488
Description
SOS1 Monoclonal specifically detects SOS1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| SOS1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GF1GGF1, GINGF, guanine nucleotide exchange factor, HGFhereditary, 1, NS4, son of sevenless homolog 1, son of sevenless homolog 1 (Drosophila), SOS-1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The SOS-01 reacts with human Sos, an ubiquitously expressed 150 kDa intracellular protein. | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| SOS-01 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Q07889 | |
| SOS1 | |
| Peptide corresponding to amino acids THPSMHRDGPPLLENAHSS of human Sos protein (NM_005633.3) | |
| 0.1 mL | |
| Breast Cancer, Cancer | |
| 6654 | |
| Protein A purified | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction