Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOS1 Antibody (SOS-01), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11068800V
Description
SOS1 Monoclonal specifically detects SOS1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SOS1 | |
Monoclonal | |
DyLight 405 | |
Q07889 | |
SOS1 | |
Peptide corresponding to amino acids THPSMHRDGPPLLENAHSS of human Sos protein (NM_005633.3) | |
0.1 mL | |
Breast Cancer, Cancer | |
6654 | |
Human, Mouse | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
SOS-01 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
GF1GGF1, GINGF, guanine nucleotide exchange factor, HGFhereditary, 1, NS4, son of sevenless homolog 1, son of sevenless homolog 1 (Drosophila), SOS-1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The SOS-01 reacts with human Sos, an ubiquitously expressed 150 kDa intracellular protein. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction