Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596239 Supplier Invitrogen™ Supplier No. PA596239
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLKSPGVRPS NPRPGTMRHP TPLQQEPRKI SYSRIPESET ESTASAPNSP RTPLTPPPAS GASSTTDVCS VFDSDHSSPF HSSNDTVFIQ VTLPHGPRSA SVSSISLTKG TDEVPVPPPV PPRRRPESAP AESSPSKIMS KHLDSPPAIP PRQPTSKAYS PRYSISDRTS ISDPPESPPL LPPREPVRTP DVFSSSPLHL QPPPLGKKSD HGNAFFPNSP SPFTPPPPQT PSPHGTRRHL PSPPLTQEVD LHSIAGPPVP PRQSTSQHIP KLPPKTYKRE HTHPSMHRDG PPLLENAHSS; Positive Samples: HeLa, Mouse liver, Rat brain, Rat liver.

Son of sevenless (SOS) is a guanine nucleotide exchange factor that activates Ras in response to growth factor stimulation. Sos1, and its closely related Sos2 paralog, are comprised of an N-terminal histone-binding domain, a RacGEF catalytic (DH) domain juxtaposed with a PH domain, a Ras exchanger motif (REM) domain, a RasGEF catalytic domain, and a C-terminal proline-rich region. Sos1 catalyzes the GDP-GTP exchange within the membrane-bound GTPase Ras and thereby switches on a key signaling circuit that involves the activation of MAPK cascade central to cellular proliferation, survival and differentiation. Growth factor stimulation rapidly induces the phosphorylation of Sos on multiple serine and threonine sites. Phosphorylation of residues within the Sos C-terminus by the MAP/ERK kinase disrupts the Grb2-Sos complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOS1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SOS1
Gene Accession No. Q07889, Q62245
Gene Alias 4430401P03Rik; 9630010N06; AI449023; G GINGF; GF1; GGF1; GINGF; gingival fibromatosis; gingival fibromatosis, hereditary, 1; guanine nucleotide exchange factor; guanine nucleotide releasing factor; HGF; NS4; ras guanine exchange factor; son of sevenless homolog 1; Son of sevenless homolog 1 (Drosophila); son-of-sevenless 1; SOS Ras/Rac guanine nucleotide exchange factor 1; SOS1; SOS-1
Gene Symbols SOS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1034-1333 of human SOS1 (NP_0056242).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20662, 313845, 6654
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.