Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOS2 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP105958IR
Description
SOS2 Polyclonal antibody specifically detects SOS2 in Human, Mouse samples. It is validated for Western BlotSpecifications
SOS2 | |
Polyclonal | |
Western Blot | |
FLJ25596, guanine nucleotide exchange factor, guanine nucleotide releasing factor, son of sevenless (Drosophilia) homolog 2, son of sevenless homolog 2, son of sevenless homolog 2 (Drosophila), SOS-2 | |
The immunogen recognized by this antibody maps to a region between residue 1282 and 1332 of human son of sevenless ortholog 2 using the numbering given in entry NP_008870.2 (GeneID 6655). | |
0.1 mL | |
Breast Cancer, Cancer | |
6655 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction