Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527492
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527492 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527492 Supplier Invitrogen™ Supplier No. PA527492
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549. Predicted reactivity: Mouse (95%), Rat (95%), Rhesus Monkey (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SOS2 promotes the exchange of Ras-bound GDP by GTP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOS2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.01% thimerosal; pH 7
Gene SOS2
Gene Accession No. Q07890
Gene Alias mSOS-2; NS9; son of sevenless 1; son of sevenless homolog 2; son of sevenless homolog 2 (Drosophila); SOS Ras/Rho guanine nucleotide exchange factor 2; Sos1; Sos2; SOS-2
Gene Symbols SOS2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SOS2. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6655
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.