Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550140 Supplier Invitrogen™ Supplier No. PA550140
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total SOS2 protein.

SOS2 promotes the exchange of Ras-bound GDP by GTP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOS2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SOS2
Gene Accession No. Q07890
Gene Alias mSOS-2; NS9; son of sevenless 1; son of sevenless homolog 2; son of sevenless homolog 2 (Drosophila); SOS Ras/Rho guanine nucleotide exchange factor 2; Sos1; Sos2; SOS-2
Gene Symbols SOS2
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human SOS2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6655
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.