Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOST/Sclerostin Antibody (2), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289575APC
Description
SOST/Sclerostin Monoclonal antibody specifically detects SOST/Sclerostin in Human samples. It is validated for ELISASpecifications
SOST/Sclerostin | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
2 | |
ELISA | |
sclerostin, VBCHsclerosteosis | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human SOST/Sclerostin (Accession#: NP_079513.1; Gln 24-Tyr 213). | |
0.1 mL | |
Signal Transduction | |
50964 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction