Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOST/Sclerostin Antibody (RM0129-4D77), Alexa Fluor™ 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122506AF350
Description
SOST/Sclerostin Monoclonal specifically detects SOST/Sclerostin in Mouse samples. It is validated for Western Blot.Specifications
SOST/Sclerostin | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
SOST | |
Mouse recombinant protein of Sclerostin | |
0.1 mL | |
Primary | |
Mouse | |
IgG2 |
Western Blot | |
RM0129-4D77 | |
Western Blot | |
sclerostin, VBCHsclerosteosis | |
Rat | |
Protein A or G purified | |
Signal Transduction | |
50964 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction