Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOST/Sclerostin Antibody (RM0129-4D77), mFluor Violet 500 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122506MFV500
Description
SOST/Sclerostin Monoclonal antibody specifically detects SOST/Sclerostin in Mouse samples. It is validated for Western BlotSpecifications
| SOST/Sclerostin | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0129-4D77 | |
| Western Blot | |
| sclerostin, VBCHsclerosteosis | |
| Mouse recombinant protein of Sclerostin | |
| 0.1 mL | |
| Signal Transduction | |
| 50964 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction