Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Sostdc1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570641
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
May be involved in the onset of endometrial receptivity for implantation/sensitization for the decidual cell reaction Enhances Wnt signaling and inhibits TGF-beta signaling. Directly antagonizes activity of BMP2, BMP4, BMP6 and BMP7 in a dose-dependent manner.
Specifications
Sostdc1 | |
Polyclonal | |
Unconjugated | |
SOSTDC1 | |
0610006G05Rik; CDA019; cystine-knot containing secreted protein; DAND7; Ectodermal BMP inhibitor; ECTODIN; sclerostin domain containing 1; sclerostin domain-containing protein 1; Sclerostin-like protein; similar to cystine knot-containing secreted protein; Sostdc1; Sostl; USAG1; USAG-1; Uterine sensitization-associated gene 1 protein; uterine sensitization-associated protein 1; uterine sensitization-associated protein-1; Wise; Wnt-signaling modulator | |
Rabbit | |
Affinity Chromatography | |
RUO | |
66042 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9CQN4 | |
SOSTDC1 | |
synthetic peptide directed towards the C-terminal region of mouse Sostdc1. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction