Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sostdc1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5121587
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121587 20 μL
PIPA5121586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121587 Supplier Invitrogen™ Supplier No. PA5121587
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

May be involved in the onset of endometrial receptivity for implantation/sensitization for the decidual cell reaction Enhances Wnt signaling and inhibits TGF-beta signaling. Directly antagonizes activity of BMP2, BMP4, BMP6 and BMP7 in a dose-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sostdc1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SOSTDC1
Gene Accession No. Q6X4U4
Gene Alias 0610006G05Rik; CDA019; cystine-knot containing secreted protein; DAND7; Ectodermal BMP inhibitor; ECTODIN; sclerostin domain containing 1; sclerostin domain-containing protein 1; Sclerostin-like protein; similar to cystine knot-containing secreted protein; Sostdc1; Sostl; USAG1; USAG-1; Uterine sensitization-associated gene 1 protein; uterine sensitization-associated protein 1; uterine sensitization-associated protein-1; Wise; Wnt-signaling modulator
Gene Symbols SOSTDC1
Host Species Rabbit
Immunogen Phe24-Ser206 (Accession Q6X4U4), with N-terminal His Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25928
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.