Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sostdc1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572000
Description
May be involved in the onset of endometrial receptivity for implantation/sensitization for the decidual cell reaction Enhances Wnt signaling and inhibits TGF-beta signaling (By similarity). Directly antagonizes activity of BMP2, BMP4, BMP6 and BMP7 in a dose-dependent manner.
Specifications
Sostdc1 | |
Polyclonal | |
Unconjugated | |
SOSTDC1 | |
Sclerostin domain-containing protein 1; Ectodermal BMP inhibitor; Ectodin; Uterine sensitization-associated gene 1 protein; USAG-1; SOSTDC1; USAG1 | |
Rabbit | |
Protein A | |
RUO | |
25928, 66042 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q6X4U4, Q9CQN4 | |
SOSTDC1 | |
KLH conjugated synthetic peptide between 169-196 amino acids from the C-terminal region of human SOSTDC1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction