Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva SOURCE™ 30S Chromatography Media

Catalog No. 45002026 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-026 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-026 Supplier Cytiva Supplier No. 17127301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemically stable cation exchanger for high resolution polishing purification of proteins, peptides and oligonucleotides

  • Average particle size: 30μm
  • Ligand: sulphonate group
  • Matrix: polystyrene/divinylbenzene
  • pH stability working range: 2 to 13
  • Available in 50mL bottle

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mL
For Use With (Application) For High Resolution Polishing PurifiCatIon of Proteins, Peptides and Oligonucleotides
Stability Stable in common Ion Exchange Buffers
Storage Requirements 4°C to 30°C
Product Type Source 30 S chromatography media
Flow Rate 2000 cm/Hoμr
Particle Size 30 μm
Height (English) Bed 3.93 in.
Height (Metric) Bed 10 cm
Max. Pressure 1000 kPa
Matrix Polystyrene/Divinylbenzene
pH Range 2 to 13
Storage Buffer 20% Ethanol, 0.2 M Sodium Acetate
Ligand Type Sulphonate Group
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.